Learn More
Glycerol, ultrapure, Spectrophotometric Grade
Description
Glycerol is used as emulsifier, thickening agent in cosmetics and foods, plasticizer in packaging, paints and paper industry, preservative in food and beverages. It is used in pharmaceutical preparations to improve smoothness and providing lubrication. It is also used in epichlorohydrin and mono propylene glycol production. It is used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 18°C |
Color | Colorless |
Density | 1.26 |
Boiling Point | 182°C (20 mm) |
Flash Point | 160°C (320°F) |
Ignition Point | 400°C (752°F) |
Odor | Odorless |
Refractive Index | 1.474 |
Quantity | 4 x 1 L |
Beilstein | 635685 |
Show More |
Safety and Handling
Hazard Category | H319: Causes serious eye irritation. |
EINECSNumber | 200-289-5 |
RTECSNumber | MA8050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.