Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Glycine, 0.2M buffer soln., pH 2.5, low endotoxin

Click to view available options
Quantity:
100mL
250mL
56-40-6
C2H5NO2
75.067
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
56-40-6
C2H5NO2
75.067
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
Concentration 0.2M buffer soln.
Physical Form Liquid
Odor Odorless
pH 2.5
Color Colorless
Quantity 100mL
Chemical Name or Material Glycine, low endotoxin