Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine N-Methyltransferase/GNMT Antibody (OTI8A3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP271838 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271838 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271838 Supplier Novus Biologicals Supplier No. NBP271838
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glycine N-Methyltransferase/GNMT Monoclonal specifically detects Glycine N-Methyltransferase/GNMT in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen Glycine N-Methyltransferase/GNMT
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone OTI8A3
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:1000, Flow Cytometry 1:100, CyTOF-ready
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias EC 2.1.1.20, glycine N-methyltransferase
Gene Symbols GNMT
Host Species Mouse
Immunogen Full length human recombinant protein of human GNMT (NP_061833) produced in HEK293T cell.
Molecular Weight of Antigen 32.6 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 27232
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.