Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine Receptor Alpha 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP226171 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226171 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226171 Supplier Novus Biologicals Supplier No. NBP226171
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Glycine Receptor Alpha 1 Polyclonal specifically detects Glycine Receptor Alpha 1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Peptide ELISA.

Specifications

Antigen Glycine Receptor Alpha 1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias GLRA 1, Glycine receptor 48 kDa subunit, Glycine receptor strychnine-binding subunit, glycine receptor subunit alpha-1, glycine receptor, alpha 1, glycine receptor, alpha 1 (startle disease/hyperekplexia), MGC138878, MGC138879, STHE
Gene Symbols GLRA1
Host Species Goat
Immunogen Peptide with sequence CLQAKDGISVKGAN corresponding to internal region according to NP_037265.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Diabetes Research, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2741
Test Specificity The immunizing peptide represents part of the cytoplasmic domain. The antibody is not expected to cross-react with other glycine receptor alpha proteins.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.