Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine Receptor Alpha 1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300113 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300113 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300113 Supplier Novus Biologicals Supplier No. NB300113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Glycine Receptor Alpha 1 Polyclonal specifically detects Glycine Receptor Alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen Glycine Receptor Alpha 1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen 1:1000
Formulation No Preservative
Gene Accession No. P07727
Gene Alias GLRA 1, Glycine receptor 48 kDa subunit, Glycine receptor strychnine-binding subunit, glycine receptor subunit alpha-1, glycine receptor, alpha 1, glycine receptor, alpha 1 (startle disease/hyperekplexia), MGC138878, MGC138879, STHE
Gene Symbols GLRA1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH. Accession # P07727
Molecular Weight of Antigen 48 kDa
Purification Method Antigen Affinity-purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Diabetes Research, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2741
Test Specificity Specific for the ∼48 kDa alpha1- and alpha2-subunits of the glycine receptor in Western blots of rat spinal cord and brain stem and in cell extracts. Does not recognize other glycine receptor subunits.
Reconstitution Reconstitute with 50 uL PBS to desired concentration.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.