Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycogen Synthase Antibody (3A7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237643
Description
Glycogen Synthase Monoclonal specifically detects Glycogen Synthase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Glycogen Synthase | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
GYS1 | |
Purified recombinant fragment of human GYS1 expressed in E. Coli. | |
Unpurified | |
RUO | |
2997 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA | |
3A7 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
EC 2.4.1.11, glycogen [starch] synthase, muscle, glycogen synthase 1 (muscle), GYSGSY | |
Mouse | |
85 kDa | |
0.1 mL | |
Primary | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction