Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycogen Synthase Antibody (3A7), Novus Biologicals™

Mouse Monoclonal Antibody
$562.50
Specifications
Antigen | Glycogen Synthase |
---|---|
Clone | 3A7 |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
Glycogen Synthase Monoclonal specifically detects Glycogen Synthase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Glycogen Synthase | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
Monoclonal | |
Ascites | |
RUO | |
Ascites with 0.03% Sodium Azide | |
2997 | |
Purified recombinant fragment of human GYS1 expressed in E. Coli. | |
Primary | |
85 kDa |
3A7 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Human | |
EC 2.4.1.11, glycogen [starch] synthase, muscle, glycogen synthase 1 (muscle), GYSGSY | |
GYS1 | |
IgG1 | |
Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title