Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycogenin 1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP17929120UL

 View more versions of this product

Catalog No. NBP1792920


Description

Description

Glycogenin 1 Polyclonal specifically detects Glycogenin 1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Glycogenin 1
Polyclonal
Western Blot 1:1000
NP_038783
GYG1
Synthetic peptide directed towards the C terminal of human Gyg. Peptide sequence SDLSFGEAPAAPQPSMSSEERKERWEQGQADYMGADSFDNIKRKLDTYLQ.
Affinity Purified
RUO
2992
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
EC 2.4.1.186, glycogenin, glycogenin 1, glycogenin-1, GN1, GN-1, GSD15, GYG
Rabbit
37 kDa
20 μL
Primary
Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.