Learn More
Glycolaldehyde diethyl acetal, stab. with ca 0.1% sodium carbonate, 98%
Description
Glycolaldehyde diethyl acetal is used in the preparation of neooxazolomycin, which is a part of the oxazolomycin family of antibiotics. It is also used in the preparation of pyrimidine based inhibitors of phosphodiesterase 7.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Glycolaldehyde diethyl acetal is used in the preparation of neooxazolomycin, which is a part of the oxazolomycin family of antibiotics. It is also used in the preparation of pyrimidine based inhibitors of phosphodiesterase 7.
Solubility
Miscible with water, chloroform and ethyl acetate.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acid and bases.
Specifications
Specifications
| Boiling Point | 166°C to 167°C |
| Flash Point | 62°C (143°F) |
| Refractive Index | 1.416 |
| Quantity | 10 g |
| Beilstein | 773727 |
| Solubility Information | Miscible with water,chloroform and ethyl acetate. |
| Formula Weight | 134.18 |
| Percent Purity | 98% |
| Chemical Name or Material | Glycolaldehyde diethyl acetal, stabilized with ≈0.1% sodium carbonate |
Safety and Handling
| Hazard Category | H227 |
| Hazard Statement | GHS H Statement H227 Combustible liquid. |
| Precautionary Statement | P210-P280-P370+P378q-P501c |
| EINECSNumber | 210-697-5 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.