Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycophorin A Antibody (GYPA/3219R), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB382260 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382260 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382260 Supplier Novus Biologicals Supplier No. NBP308261DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Glycophorin A Monoclonal antibody specifically detects Glycophorin A in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Glycophorin A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GYPA/3219R
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR
Host Species Rabbit
Immunogen Recombinant full-length human Glycophorin A protein (Uniprot: P02724)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2993
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.