Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycophorin A Antibody (M2A1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB016240 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016240 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016240 Supplier Novus Biologicals Supplier No. NBP281107AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Glycophorin A Monoclonal specifically detects Glycophorin A in Human samples. It is validated for ELISA, Immunoblotting.

Specifications

Antigen Glycophorin A
Applications ELISA, Immunoblot
Classification Monoclonal
Clone M2A1
Conjugate Alexa Fluor 532
Dilution ELISA, Immunoblotting
Formulation 50mM Sodium Borate
Gene Alias CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR
Gene Symbols GYPA
Host Species Rabbit
Immunogen M2A1 was prepared by immunzing BALB/c mice with human red blood cells of the M phenotype, and was screened by testing for anti-M activity.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2993
Test Specificity M2A1 binds specifically to the M antigen on human erythrocytes, and shows no reactivity to N antigen. The epitope for this antibody includes the amino-terminal serine residue and sialic acid residues of glycophorin A. The reactivity of M2A1 for M antigen is pH- and salt-dependent (optimum ∽ pH8-9). Blood group antigen M is located on the major sialoglycoprotein (glycophorin A) of red blood cells.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.