Learn More
Description
Specifications
Specifications
| Antigen | Glycophorin A |
| Applications | ELISA, Immunoblot |
| Classification | Monoclonal |
| Clone | M2A1 |
| Conjugate | DyLight 594 |
| Dilution | ELISA, Immunoblotting |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR |
| Gene Symbols | GYPA |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
