Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycophorin A Antibody (MM0314-7S41), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP212339Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212339Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212339Z Supplier Novus Biologicals Supplier No. NBP212339AF700

Mouse Monoclonal Antibody

Glycophorin A Monoclonal specifically detects Glycophorin A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Glycophorin A
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MM0314-7S41
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. AAA52768.1
Gene Alias CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR
Gene Symbols GYPA
Host Species Mouse
Immunogen Human recombinant Glycophorin A (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2993
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.