Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycophorin A Antibody (SPM599) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP247719B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247719B 0.2 mg
NBP247719A 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP247719B Supplier Novus Biologicals Supplier No. NBP2477190.2MG

Mouse Monoclonal Antibody

Glycophorin A Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.

Specifications

Antigen Glycophorin A
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Classification Monoclonal
Clone SPM599
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.25 - 0.5 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR
Gene Symbols GYPA
Host Species Mouse
Immunogen Recombinant human glycophorin A protein
Molecular Weight of Antigen 39 kDa
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2993
Test Specificity Recognizes a sialoglycoprotein of 39kDa, identified as glycophorin A (GPA). It is present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorin acts as the receptor for Sandei virus and parvovirus. Glycophorins A (GPA) and B (GPB), which are single, trans-membrane sialoglycoproteins. GPA is the carrier of blood group M and N specificities, while GPB accounts for S and U specificities. GPA and GPB provide the cells with a large mucin like surface and it has been suggested this provides a barrier to cell fusion, so minimizing aggregation between red blood cells in the circulation.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.