Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycophorin C Antibody (BRIC10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N106622A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106622A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106622A30 Supplier Novus Biologicals Supplier No. NB10066522AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glycophorin C Monoclonal specifically detects Glycophorin C in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen Glycophorin C
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone BRIC10
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2′, Sialoglycoprotein D
Gene Symbols GYPC
Host Species Mouse
Immunogen Human erythrocytes
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2995
Test Specificity NB100-66522 recognizes an epitope in the N-terminal region of CD236, also known as glycophorin C (GPC). CD236 is an integral membrane protein which carries the Gerbich blood group antigens. Expression of CD236 is not restricted to erythroid cells, but is broadly distributed in both erythroid and non-erythroid tissues.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.