Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycoprotein V/CD42d Antibody (1C2), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N2071PET594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N2071PET594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N2071PET594 Supplier Novus Biologicals Supplier No. NBP200071PEATT594
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Glycoprotein V/CD42d Monoclonal specifically detects Glycoprotein V/CD42d in Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Glycoprotein V/CD42d
Applications Flow Cytometry
Classification Monoclonal
Clone 1C2
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias platelet glycoprotein V, CD42d, glycoprotein V (platelet), GPV
Gene Symbols GP5
Host Species Armenian Hamster
Immunogen The immunogen for this antibody is CD42d (platelet glycoprotein V).
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2814.0
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.