Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycoprotein V/CD42d Antibody (508209), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB370486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370486 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370486 Supplier Novus Biologicals Supplier No. FAB4249W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glycoprotein V/CD42d Monoclonal specifically detects Glycoprotein V/CD42d in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Glycoprotein V/CD42d
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 508209
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias platelet glycoprotein V, CD42d, glycoprotein V (platelet), GPV
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Glycoprotein V/CD42d, Gln17-Gly523, Accession # P40197
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2814
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.