Learn More
Glyoxal dimethyl acetal, 60% aq. soln.
Description
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.
Solubility
Fully miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.15 |
| Boiling Point | 100°C to 102°C |
| Flash Point | 102°C (216°F) |
| Refractive Index | 1.414 |
| Quantity | 2000 g |
| Beilstein | 1850744 |
| Solubility Information | Fully miscible in water. |
| Formula Weight | 104.11 |
| Concentration or Composition (by Analyte or Components) | 60% aq. soln. |
| Chemical Name or Material | Glyoxal dimethyl acetal |
Safety and Handling
| Hazard Category | H317 |
| Hazard Statement | GHS H Statement H317 May cause an allergic skin reaction. |
| Precautionary Statement | P261-P272-P280g-P302+P352-P333+P313-P363-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.