Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyoxalase I Antibody (Glo1a), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB338543 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338543 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338543 Supplier Novus Biologicals Supplier No. NBP119015MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glyoxalase I Monoclonal antibody specifically detects Glyoxalase I in Human, Mouse samples. It is validated for Western Blot, Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Glyoxalase I
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone Glo1a
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Simple Western, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase
Host Species Mouse
Immunogen Full length human GLO1 protein [Swiss-Prot #Q04760].
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2739
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.