Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glypican 1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB119967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119967 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119967 Supplier Novus Biologicals Supplier No. NBP142718UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glypican 1 Polyclonal specifically detects Glypican 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Glypican 1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1
Host Species Rabbit
Immunogen Glypican 1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human glypican-1 protein. (Uniprot: P35052)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2817
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.