Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glypican 3 Antibody (307801), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB349245 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349245 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349245 Supplier Novus Biologicals Supplier No. FAB2119MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glypican 3 Monoclonal specifically detects Glypican 3 in Human, Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot.

Specifications

Antigen Glypican 3
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 307801
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Glypican 3, Gln25-Val558, Accession # P51654.1
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2719
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.