Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glypican 4 Antibody (51E3), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N249892PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N249892PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N249892PEC5 Supplier Novus Biologicals Supplier No. NBP249892PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glypican 4 Monoclonal specifically detects Glypican 4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Glypican 4
Applications Flow Cytometry
Classification Monoclonal
Clone 51000
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias glypican 4, glypican proteoglycan 4, glypican-4, K-glypicandJ900E8.1 (glypican 4)
Gene Symbols GPC4
Host Species Mouse
Immunogen Recombinant human GPC4(401-529aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2239.0
Test Specificity Anti-human GPC4 mAb, clone AT51E3, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human GPC4 protein.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.