Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Antibody (6804) [DyLight 405], Novus Biologicals™
SDP

Catalog No. NB376973 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376973 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376973 Supplier Novus Biologicals Supplier No. FAB615E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry (Intracellular Staining).

Specifications

Antigen GM-CSF
Applications CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 6804
Conjugate DyLight 405
Dilution ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Host Species Mouse
Immunogen E. coli-derived recombinant human GM-CSF, Ala18-Glu144, Accession # P04141
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1437
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.