Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Antibody, Novus Biologicals™
SDP

Catalog No. NB600632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600632 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600632 Supplier Novus Biologicals Supplier No. NB600632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

GM-CSF Polyclonal specifically detects GM-CSF in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen GM-CSF
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:1000-1:5000, Immunohistochemistry 1:10-1:500
Formulation Antiserum with No Preservative
Gene Accession No. P04141
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Gene Symbols CSF2
Host Species Rabbit
Immunogen This whole rabbit serum was prepared by repeated immunizations with full length recombinant human GM-CSF. (Uniprot: P04141)
Molecular Weight of Antigen 16.29 kDa
Purification Method Unpurified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1437
Test Specificity In ELISA and other immunoreactive assays, this antiserum will recognize both native and recombinant human GM-CSF in cell supernatants and certain body fluids.
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.