Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Antibody (OTI8G5), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP270462A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270462A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270462A30 Supplier Novus Biologicals Supplier No. NBP270462AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen GM-CSF
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI8G5
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Gene Symbols CSF2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 18-144 of human GM-CSF (NP_000749) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1437
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.