Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Antibody (SPM469) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB397109 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397109 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397109 Supplier Novus Biologicals Supplier No. NBP2347740.1MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

GM-CSF Monoclonal antibody specifically detects GM-CSF in Human, Cynomolgus Monkey, Macaque samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF

Specifications

Antigen GM-CSF
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone SPM469
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5-1 μg/million cells, Immunocytochemistry/ Immunofluorescence 0.5-1 μg/mL, CyTOF-ready
Formulation 10 mM PBS
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Host Species Rat
Immunogen Recombinant human GM-CSF protein (Uniprot: P04141)
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1437
Target Species Human, Cynomolgus Monkey, Macaque
Content And Storage Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.