Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSFR alpha Antibody (4H1), Novus Biologicals™
SDP

Catalog No. NB018538 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018538 25 μg
NBP14336101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018538 Supplier Novus Biologicals Supplier No. NBP14336125UG

Mouse Monoclonal Antibody

GM-CSFR alpha Monoclonal specifically detects GM-CSFR alpha in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GM-CSF R alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 4H1
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000
Formulation Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Gene Alias CD116, CD116 antigen, CDw116, colony stimulating factor 2 receptor alpha subunit, colony stimulating factor 2 receptor, alpha, low-affinity(granulocyte-macrophage), CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF receptor alpha subunit, GMCSFR, GM-CSF-R-alpha, GMCSFR-alpha, GMR, GMR-alpha, granulocyte-macrophage colony-stimulating factor receptor alpha chain, granulocyte-macrophage colony-stimulating factor receptor subunit alpha, MGC3848, MGC4838
Gene Symbols CSF2RA
Host Species Mouse
Immunogen The immunogen for this antibody was CD116.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1438
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.