Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GM-CSFR alpha Antibody (4H1), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | GM-CSF R alpha |
---|---|
Clone | 4H1 |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018538
![]() |
Novus Biologicals
NBP14336125UG |
25 μg |
Each for $206.50
|
|
|||||
NBP14336101
![]() |
Novus Biologicals
NBP1433610.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
Description
GM-CSFR alpha Monoclonal specifically detects GM-CSFR alpha in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
GM-CSF R alpha | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Mouse | |
Cancer | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
1438 | |
The immunogen for this antibody was CD116. | |
Primary | |
Store at 4C. Do not freeze. |
4H1 | |
Flow Cytometry | |
Unconjugated | |
RUO | |
Human | |
CD116, CD116 antigen, CDw116, colony stimulating factor 2 receptor alpha subunit, colony stimulating factor 2 receptor, alpha, low-affinity(granulocyte-macrophage), CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF receptor alpha subunit, GMCSFR, GM-CSF-R-alpha, GMCSFR-alpha, GMR, GMR-alpha, granulocyte-macrophage colony-stimulating factor receptor alpha chain, granulocyte-macrophage colony-stimulating factor receptor subunit alpha, MGC3848, MGC4838 | |
CSF2RA | |
IgG1 κ | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title