Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSFR alpha Antibody (698423), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB377019 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377019 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377019 Supplier Novus Biologicals Supplier No. FAB6130D
Add to Cart
Add to Cart

Rat Monoclonal Antibody

GM-CSFR alpha Monoclonal specifically detects GM-CSFR alpha in Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Neutralization.

Specifications

Antigen GM-CSFR alpha
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Neutralization
Classification Monoclonal
Clone 698423
Conjugate DyLight 350
Dilution Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD116, CD116 antigen, CDw116, colony stimulating factor 2 receptor alpha subunit, colony stimulating factor 2 receptor, alpha, low-affinity(granulocyte-macrophage), CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF receptor alpha subunit, GMCSFR, GM-CSF-R-alpha, GMCSFR-alpha, GMR, GMR-alpha, granulocyte-macrophage colony-stimulating factor receptor alpha chain, granulocyte-macrophage colony-stimulating factor receptor subunit alpha, MGC3848, MGC4838
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GM-CSF R alpha , Leu30-Pro327, Accession # Q00941
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1438
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.