Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSFR alpha Rabbit anti-Human, Clone: 9, Novus Biologicals™
SDP

Catalog No. NB020704 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020704 50 μg
NB020703 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020704 Supplier Novus Biologicals Supplier No. NBP28961050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GM-CSFR alpha Monoclonal specifically detects GM-CSFR alpha in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen GM-CSFR alpha
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 9
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias CD116, CD116 antigen, CDw116, colony stimulating factor 2 receptor alpha subunit, colony stimulating factor 2 receptor, alpha, low-affinity(granulocyte-macrophage), CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF receptor alpha subunit, GMCSFR, GM-CSF-R-alpha, GMCSFR-alpha, GMR, GMR-alpha, granulocyte-macrophage colony-stimulating factor receptor alpha chain, granulocyte-macrophage colony-stimulating factor receptor subunit alpha, MGC3848, MGC4838
Host Species Rabbit
Immunogen Recombinant Human GM-CSFR alpha protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1438
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.