Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM130/GOLGA2 Antibody (2059D) - Azide and BSA Free, R&D Systems™
SDP

Catalog No. MAB8199 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8199 100 μg
MAB8199SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB8199 Supplier Novus Biologicals Supplier No. MAB8199
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GM130/GOLGA2 Monoclonal specifically detects GM130/GOLGA2 in Human samples. It is validated for ICC/IF (Negative), Immunohistochemistry (Negative), Western Blot.

Specifications

Antigen GM130/GOLGA2
Applications ICC/IF (Negative), Immunohistochemistry (Negative), Western Blot
Classification Monoclonal
Clone 2059D
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry (Negative), ICC/IF (Negative)
Formulation PBS with No Preservative
Gene Alias 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein
Gene Symbols GOLGA2
Host Species Rabbit
Immunogen Partial recombinant human GOLG-A2 protein (amino acids 528-606) [UniProt Q08379]
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2801
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.