Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM130/GOLGA2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP253420SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP253420SS 0.025 mg
NBP253420 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP253420SS Supplier Novus Biologicals Supplier No. NBP253420SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

GM130/GOLGA2 Polyclonal specifically detects GM130/GOLGA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GM130/GOLGA2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, Immunohistochemistry 1:1000 - 1:1500, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:1500
Formulation PBS with 0.02% Sodium Azide
Gene Alias 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein
Gene Symbols GOLGA2
Host Species Rabbit
Immunogen Partial recombinant human GM130/GOLGA2 protein (amino acids 528-606). [UniPro Q08379]
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2801
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.