Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM130/GOLGA2 Antibody (NN 2C10/1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB1002622AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002622AG 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002622AG Supplier Novus Biologicals Supplier No. NB1002622AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GM130/GOLGA2 Monoclonal specifically detects GM130/GOLGA2 in Rat samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen GM130/GOLGA2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone NN 2C10/1
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein
Gene Symbols GOLGA2
Host Species Mouse
Immunogen Recombinant purified rat GM130/GOLGA2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2801
Test Specificity Does not cross react with human protein.
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.