Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM2 Ganglioside Antibody (L55-81), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB016959 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016959 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016959 Supplier Novus Biologicals Supplier No. NBP281278JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GM2 Ganglioside Monoclonal specifically detects GM2 Ganglioside in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GM2 Ganglioside
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone L55-81
Conjugate Janelia Fluor 646
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias GM2 (ganglioside), Tay-Sachs ganglioside
Host Species Rabbit
Immunogen L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanoma.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity L55-81 binds specifically to GM2 ganglioside; this antibody shows no reactivity towards GM1, GM3, GD2 or normal tissues- the L55-81 epitope contains both a GalNAc and sialic acid bound to lactose. Ganglioside GM2 (GalNAcbeta1-4(NeuNAcalpha2-3)Galbeta1-4Glcbeta-1ceramide) is a glycosphingolipid present on the cell surface and a major tumor-differentiation antigen in human melanoma and hepatoma.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.