Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMPPB Antibody, Novus Biologicals™
SDP

Catalog No. NBP216695 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216695 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216695 Supplier Novus Biologicals Supplier No. NBP216695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GMPPB Polyclonal specifically detects GMPPB in Human samples. It is validated for Western Blot.

Specifications

Antigen GMPPB
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9Y5P6
Gene Alias EC 2.7.7.13, GDP-mannose pyrophosphorylase BKIAA1851, GTP-mannose-1-phosphate guanylyltransferase beta, mannose-1-phosphate guanyltransferase beta, mannose-1-phosphate guanylyltransferase
Gene Symbols GMPPB
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GMPPB. The exact sequence is proprietary.
Molecular Weight of Antigen 40 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29925
Test Specificity Human. Expected to cross react based on sequence identity: Mouse (99%), Rat (99%), Zebrafish (81%), Xenopus tropicalis (83%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.