Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMPR2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB330287 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB330287 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB330287 Supplier Novus Biologicals Supplier No. NBP297734PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GMPR2 Polyclonal antibody specifically detects GMPR2 in Human samples. It is validated for Western Blot

Specifications

Antigen GMPR2
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias EC 1.7.1.7, GMP reductase 2, Guanosine 5'-monophosphate oxidoreductase 2, guanosine monophosphate reductase 2MGC830, guanosine monophosphate reductase isolog, MGC15084
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GMPR2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 51292
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.