Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNAQ Antibody (GNAQ/2434) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119311 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119311 Supplier Novus Biologicals Supplier No. NBP308585
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GNAQ Monoclonal specifically detects GNAQ in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GNAQ
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone GNAQ/2434
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias GAQG-ALPHA-q, guanine nucleotide binding protein (G protein), q polypeptide, Guanine nucleotide-binding protein alpha-q, guanine nucleotide-binding protein G(q) subunit alpha
Host Species Mouse
Immunogen Recombinant full-length human GNAQ protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2776
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.