Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNB1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15530720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15530720 20 μL
NBP155307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15530720 Supplier Novus Biologicals Supplier No. NBP15530720UL

Rabbit Polyclonal Antibody

GNB1 Polyclonal specifically detects GNB1 in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen GNB1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P62873
Gene Alias beta subunit, signal-transducing proteins GS/GI, G protein, beta-1 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 1, guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 1, guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1, Transducin beta chain 1
Gene Symbols GNB1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to GNB1(guanine nucleotide binding protein (G protein), beta polypeptide 1) The peptide sequence was selected from the N terminal of GNB1. Peptide sequence MSELDQLRQEAEQLKNQIRDARKACADATLSQITNNIDPVGRIQMRTRRT
Molecular Weight of Antigen 37 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2782
Test Specificity This product is specific to Subunit or Isoform: beta-1.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.