Learn More
Description
Specifications
Specifications
| Antigen | GNB1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | P62873 |
| Gene Alias | beta subunit, signal-transducing proteins GS/GI, G protein, beta-1 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 1, guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 1, guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1, Transducin beta chain 1 |
| Gene Symbols | GNB1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
