Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNG13 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB064321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064321 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064321 Supplier Novus Biologicals Supplier No. NBP300086AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GNG13 Polyclonal specifically detects GNG13 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GNG13
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias G gamma subunit, clone:h2-35, G(gamma)13, guanine nucleotide binding protein (G protein), gamma 13, guanine nucleotide binding protein 13, gamma, guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-13, h2-35
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant GNG13 (Q9P2W3; Met1-Cys64).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51764
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.