Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNPDA1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224653SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224653SS 0.025 mg
NBP224653 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224653SS Supplier Novus Biologicals Supplier No. NBP224653SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GNPDA1 Polyclonal specifically detects GNPDA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GNPDA1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1.0 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P46926
Gene Alias EC 3.5.99.6, GlcN6P deaminase 1, glucosamine-6-phosphate deaminase 1KIAA0060HLNGNPIGNPDA, glucosamine-6-phosphate isomerase, glucosamine-6-phosphate isomerase 1, GNP1, GNPDA 1, GPI, oscillin
Gene Symbols GNPDA1
Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human GNPDA1 was used as the immunogen for the antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10007
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, 94% homologous in cow, 88% homologous in mouse, chicken, zebrafish, xenopus and 83% homologous in pufferfish.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.