Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNPTG Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB047988 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047988 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047988 Supplier Novus Biologicals Supplier No. NBP297807F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GNPTG Polyclonal specifically detects GNPTG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GNPTG
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias C16orf27, c316G12.3, CAB56184, GlcNAc-1-phosphotransferase subunit gamma, GNPTAGchromosome 16 open reading frame 27, LP2537, N-acetylglucosamine-1-phosphate transferase, gamma subunit, N-acetylglucosamine-1-phosphotransferase subunit gamma, N-acetylglucosamine-1-phosphotransferase, gamma subunit, RJD9, UDP-N-acetylglucosamine-1-phosphotransferase subunit gamma
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GNPTG fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84572
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.