Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GnRHR Antibody (GNRHR/2982R), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB384552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384552 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384552 Supplier Novus Biologicals Supplier No. NBP308770AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GnRHR Monoclonal antibody specifically detects GnRHR in Human samples. It is validated for ELISA

Specifications

Antigen GnRHR
Applications ELISA
Classification Monoclonal
Clone GNRHR/2982R
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias GnRH receptor, GnRH-R, GNRHR1, gonadotropin-releasing hormone (type 1) receptor 1, gonadotropin-releasing hormone receptor, GRHR, leutinizing-releasing hormone receptor, LHRHRleutinizing hormone releasing horomone receptor, LRHR, luliberin receptor, type I GnRH receptor
Host Species Rabbit
Immunogen Recombinant full-length human GnRHR protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2798
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.