Learn More
GeneBLAzer™ GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 Cells
Description
- Accession Number: NM_000406.2
- Population Status: Single Cell Clone Origin
- GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 cells contain the human gonadotropin-releasing hormone receptor (GnRHR), (Accession # NM_000406.2) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 (Cat No. K1854A) cells contain a beta-lactamase reporter gene under control of the NFAT Response Element. GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of Luteinizing Hormone-Releasing Hormone (LH-RH). In addition, GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
Specifications
Specifications
| Accession Number | NM_000406.2 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 cells are shipped on dry ice, 8 x 10^6 cells⁄vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Detection Method | Fluorescence |
| For Use With (Application) | FRET |
| Human Protein Class | G-Protein Coupled Receptor (GPCR) |
| Population Status | Single Cell Clone Origin |
| Product Line | GeneBLAzer |
| Quantity | 1 vial |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.