Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat IgG Antibody (GT175), PE, Novus Biologicals™
SDP

Catalog No. NB309438 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309438 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309438 Supplier Novus Biologicals Supplier No. NBP250413PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Goat IgG Monoclonal antibody specifically detects Goat IgG in Goat samples. It is validated for Immunohistochemistry

Specifications

Antigen Goat IgG
Applications Immunohistochemistry
Classification Monoclonal
Clone GT175
Conjugate PE
Dilution Immunohistochemistry
Formulation PBS
Gene Alias Immunoglobulin G
Host Species Mouse
Immunogen Goat immunoglobulins
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Goat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.