Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GOAT/MBOAT4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25852225UL
Description
GOAT/MBOAT4 Polyclonal specifically detects GOAT/MBOAT4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GOAT/MBOAT4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
EC 2.3.1.-, ghrelin O-acyltransferase, GOATFKSG89, membrane bound O-acyltransferase domain containing 4, Membrane-bound O-acyltransferase domain-containing protein 4, OACT4, O-acyltransferase (membrane bound) domain containing 4, O-acyltransferase domain-containing protein 4 | |
Rabbit | |
Affinity Purified | |
RUO | |
619373 | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
MBOAT4 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LDDSLLHAAGFGPELGQSPGEEGYVPDADIWTLERTHRISVFSRKWNQSTARWLRRLVFQHS | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction