Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Goat Progesterone ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Click to view available options
Quantity:
1 Kit
Description
The Goat Progesterone ELISA Kit (Colorimetric) measures goat Progesterone in serum, plasma, tissue homogenates.
Specifications
Specifications
| Assay Range | 1 to 30ng/mL |
| Assay Sensitivity | 0.2ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plasma, Serum, Tissue Homogenates |
| Target Species | Goat |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Research Discipline | Signal Transduction |
| Sample Volume | 50 to 100 μL |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction