Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GODZ Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB135631 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB135631 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB135631 Supplier Novus Biologicals Supplier No. NBP306315AF700

Rabbit Polyclonal Antibody

GODZ Polyclonal specifically detects GODZ in Human samples. It is validated for Western Blot.

Specifications

Antigen GODZ
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DHHC domain containing 3, DKFZp313D2314, EC 2.3.1, EC 2.3.1.-, FLJ20209, FLJ45940, Protein DHHC1, ZDHC3, zinc finger, DHHC-type containing 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GODZ fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51304
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.