Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold(I) chloride, 99.9% (metals basis)

Catalog No. aa4336503
Click to view available options
Quantity:
1 g
5 g

CAS: 10294-29-8 | AuCl | 232.42 g/mol

Gold(I) chloride is used as a starting material for the preparation of Phosphine ligated gold(I) complexes, which is used as catalysts in synthetic chemistry. It is also used to prepare metallic gold and potassium auric bromide and potassium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-29-8
Molecular Formula AuCl
Molecular Weight (g/mol) 232.42
MDL Number MFCD00046175
InChI Key FDWREHZXQUYJFJ-UHFFFAOYSA-M
Synonym gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade
PubChem CID 27366
ChEBI CHEBI:30078
IUPAC Name chlorogold
SMILES [Cl-].[Au+]

Specifications

Melting Point 289°C (decomposition)
Physical Form Powder
Quantity 1 g
UN Number UN3260
Merck Index 14,4515
Solubility Information Soluble in hydrochloric acid,hydrobromic acid and organic solvents. Slightly soluble in water.
Formula Weight 232.42
Percent Purity 99.9%
Odor Odorless
Sensitivity Moisture sensitive
Density 7.57 g/mL
Assay (metals basis)
Chemical Name or Material Gold (I) chloride
Show More Show Less
Hazard Category H290-H314-H317-H500
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P234-P260-P264b-P272-P280-P301+P330+P331-P302+P352-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 233-655-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.