Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis)
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Gold(I) sodium thiosulfate hydrate is useful in organogold compounds. Further, it serves as an antirheumatic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.9% (Metal basis) |
| Solubility Information | Soluble in water. Insoluble in ethanol and most other organic solvents. |
| Formula Weight | 490.19 (anhydrous) |
| Appearance | White powder |
| Density | 3.09 |
| Chemical Name or Material | Gold(I) sodium thiosulfate hydrate |
Safety and Handling
| EINECSNumber | 239-324-4 |
| RTECSNumber | CK6235000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction